Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHG7
Entrez Gene ID
102177499
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Gene Aliases
-
Chromosome Location
Chr.5: 22348784 - 22417546 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049016.1 XP_017904505.1 6 - 7 670 66

View Full Product Details