Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLRA3
Entrez Gene ID
102185743
Gene Name
glycine receptor alpha 3
Gene Aliases
-
Chromosome Location
Chr.8: 6895534 - 7071416 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052791.1 XP_017908280.1 4 - 5 508 91
XM_018052792.1 XP_017908281.1 4 - 5 493 91

View Full Product Details