Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKAIN2
Entrez Gene ID
102183327
Gene Name
sodium/potassium transporting ATPase interacting 2
Gene Aliases
-
Chromosome Location
Chr.9: 13605924 - 14790661 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684494.3 XP_005684551.1 4 - 5 829 68
GenBank mRNA JT736480.1 - 7 - 8 2408 68

View Full Product Details