Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIC3
Entrez Gene ID
102184169
Gene Name
RIC3 acetylcholine receptor chaperone
Gene Aliases
-
Chromosome Location
Chr.15: 37958883 - 38017633 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059497.1 XP_017914986.1 2 - 3 379 97
XM_018059498.1 XP_017914987.1 2 - 3 379 97

View Full Product Details