Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102185949
Entrez Gene ID
102185949
Gene Name
olfactory receptor-like protein DTMT
Gene Aliases
-
Chromosome Location
Chr.19: 23990203 - 23991314 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693406.2 XP_005693463.1 1 - 1 119 65

View Full Product Details