Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C25H7orf26
Entrez Gene ID
102177492
Gene Name
chromosome 25 C7orf26 homolog
Gene Aliases
-
Chromosome Location
Chr.25: 39280997 - 39292788 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040906.1 XP_017896395.1 3 - 4 958 64
GenBank mRNA JT671358.1 - 3 - 4 522 64
KA350829.1 - 3 - 4 687 64

View Full Product Details