Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDBF2
Entrez Gene ID
102174345
Gene Name
zinc finger DBF-type containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 41304926 - 41315246 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066546.1 XP_017922035.1 2 - 3 187 84

View Full Product Details