Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOLR3
Entrez Gene ID
102174322
Gene Name
folate receptor alpha
Gene Aliases
-
Chromosome Location
Chr.15: 31639378 - 31643975 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689888.3 XP_005689945.1 2 - 3 286 58
XM_005689889.2 XP_005689946.1 2 - 3 296 58
GenBank mRNA JO520649.2 - 2 - 3 264 58

View Full Product Details