Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RILPL1
Entrez Gene ID
102182526
Gene Name
Rab interacting lysosomal protein like 1
Gene Aliases
-
Chromosome Location
Chr.17: 19061843 - 19104800 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691359.3 XP_005691416.3 2 - 3 711 67
XM_013970618.2 XP_013826072.2 2 - 3 715 67
XM_013970619.2 XP_013826073.2 2 - 3 716 67
GenBank mRNA JO427268.1 - - 1520 67

View Full Product Details