Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLEC
Entrez Gene ID
102173958
Gene Name
malectin
Gene Aliases
-
Chromosome Location
Chr.17: 8178936 - 8194129 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691571.3 XP_005691628.1 3 - 4 727 79
GenBank mRNA JO630382.2 - 3 - 4 734 79

View Full Product Details