Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CMC1
Entrez Gene ID
102174072
Gene Name
C-X9-C motif containing 1
Gene Aliases
-
Chromosome Location
Chr.22: 2582219 - 2664522 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018067031.1 XP_017922520.1 3 - 4 357 100

View Full Product Details