Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGN5
Entrez Gene ID
102181761
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 354219 - 363544 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700588.3 XP_005700645.1 2 - 3 181 68
XM_005700589.3 XP_005700646.1 2 - 3 230 68

View Full Product Details