Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAMP3
Entrez Gene ID
102175362
Gene Name
receptor activity modifying protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 43686025 - 43714339 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963384.2 XP_013818838.2 2 - 3 227 68
GenBank mRNA JT732295.1 - 5 - 6 1039 68

View Full Product Details