Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM79
Entrez Gene ID
102177674
Gene Name
transmembrane protein 79
Gene Aliases
-
Chromosome Location
Chr.3: 105608453 - 105614876 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677351.2 XP_005677408.1 2 - 3 929 110

View Full Product Details