Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8H9orf64
Entrez Gene ID
102178274
Gene Name
chromosome 8 C9orf64 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 76983173 - 76995043 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684108.3 XP_005684165.1 1 - 2 586 68

View Full Product Details