Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102168470
Entrez Gene ID
102168470
Gene Name
uncharacterized protein C8orf76
Gene Aliases
C14H8orf76
Chromosome Location
Chr.14: 65441953 - 65457242 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688867.3 XP_005688924.2 5 - 6 1345 93

View Full Product Details