Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUC15
Entrez Gene ID
102178191
Gene Name
mucin 15, cell surface associated
Gene Aliases
-
Chromosome Location
Chr.15: 25673591 - 25686762 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690015.3 XP_005690072.2 2 - 3 950 96
GenBank mRNA JO426280.1 - 2 - 3 2292 96

View Full Product Details