Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFX
Entrez Gene ID
102181669
Gene Name
zinc finger protein X-linked
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 45877350 - 45913093 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043809.1 XP_017899298.1 2 - 3 624 86
XM_018043810.1 XP_017899299.1 2 - 3 587 86
XM_018043812.1 XP_017899301.1 2 - 3 398 86
XM_018043813.1 XP_017899302.1 2 - 3 552 86
XM_018043814.1 XP_017899303.1 2 - 3 624 86
XM_018043816.1 XP_017899305.1 2 - 3 625 86

View Full Product Details