Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND2
Entrez Gene ID
102180206
Gene Name
BEN domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 52204640 - 52259227 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043708.1 XP_017899197.1 15 - 16 2241 103

View Full Product Details