Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP13
Entrez Gene ID
102170726
Gene Name
NLR family pyrin domain containing 13
Gene Aliases
-
Chromosome Location
Chr.18: 64951585 - 64972247 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063246.1 XP_017918735.1 8 - 9 2673 93

View Full Product Details