Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RWDD2B
Entrez Gene ID
102185502
Gene Name
RWD domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.1: 5527253 - 5540773 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674719.3 XP_005674776.1 5 - 6 1605 98
GenBank mRNA JO415037.2 - 1 - 2 102 98
JO434715.2 - 1 - 2 361 98
JO523644.2 - 3 - 4 512 98
JO586011.2 - 5 - 6 986 98
JT658637.1 - 5 - 6 925 98
KA342963.1 - 5 - 6 1103 98

View Full Product Details