Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H16orf70
Entrez Gene ID
102178654
Gene Name
chromosome 18 C16orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 36027843 - 36059226 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692120.3 XP_005692177.1 12 - 13 1104 98
XM_005692121.3 XP_005692178.1 12 - 13 1090 98
XM_018062103.1 XP_017917592.1 12 - 13 1092 98

View Full Product Details