Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H16orf70
Entrez Gene ID
102178654
Gene Name
chromosome 18 C16orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 36027843 - 36059226 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692120.3 XP_005692177.1 6 - 7 494 84
XM_005692121.3 XP_005692178.1 6 - 7 480 84
XM_018062103.1 XP_017917592.1 6 - 7 482 84
GenBank mRNA JO587830.2 - 5 - 6 455 84

View Full Product Details