Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCA1B
Entrez Gene ID
102191747
Gene Name
guanylate cyclase activator 1B
Gene Aliases
-
Chromosome Location
Chr.23: 32891978 - 32904510 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696310.3 XP_005696367.1 2 - 3 636 75

View Full Product Details