Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRSN1
Entrez Gene ID
102170285
Gene Name
neurensin 1
Gene Aliases
-
Chromosome Location
Chr.18: 391530 - 411569 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696787.3 XP_005696844.2 3 - 4 467 89

View Full Product Details