Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG12
Entrez Gene ID
102181760
Gene Name
ALG12 alpha-1,6-mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 118274410 - 118279543 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048932.1 XP_017904421.1 2 - 3 453 94
GenBank mRNA JO337979.2 - 4 - 5 576 94
JO640929.2 - 2 - 3 565 94

View Full Product Details