Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB3
Entrez Gene ID
102169421
Gene Name
heat shock protein family B (small) member 3
Gene Aliases
-
Chromosome Location
Chr.20: 24689780 - 24700032 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065796.1 XP_017921285.1 1 - 2 664 74
GenBank mRNA JO376428.2 - 1 - 2 628 74
JO552024.2 - 1 - 2 659 74

View Full Product Details