Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF48
Entrez Gene ID
100861031
Gene Name
zinc finger protein 48
Gene Aliases
-
Chromosome Location
Chr.25: 26489118 - 26493881 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040291.1 XP_017895780.1 2 - 3 319 73
GenBank mRNA JI861697.1 - 1 - 2 81 73

View Full Product Details