Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP22
Entrez Gene ID
102180376
Gene Name
Rho GTPase activating protein 22
Gene Aliases
-
Chromosome Location
Chr.28: 2751852 - 2933544 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042511.1 XP_017898000.1 6 - 7 875 75
XM_018042512.1 XP_017898001.1 4 - 5 850 75
XM_018042515.1 XP_017898004.1 2 - 3 531 75

View Full Product Details