Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SVIP
Entrez Gene ID
102184318
Gene Name
small VCP interacting protein
Gene Aliases
-
Chromosome Location
Chr.29: 21404618 - 21413413 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043550.1 XP_017899039.1 2 - 3 211 71

View Full Product Details