Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHNAK
Entrez Gene ID
102181273
Gene Name
AHNAK nucleoprotein
Gene Aliases
-
Chromosome Location
Chr.29: 41143144 - 41238053 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042689.1 XP_017898178.1 4 - 5 641 62
GenBank mRNA KA347340.1 - 4 - 5 614 62

View Full Product Details