Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C25H16orf71
Entrez Gene ID
102185623
Gene Name
chromosome 25 C16orf71 homolog
Gene Aliases
-
Chromosome Location
Chr.25: 3733463 - 3742719 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697485.3 XP_005697542.2 2 - 3 602 69
XM_005697486.3 XP_005697543.2 2 - 3 602 69

View Full Product Details