Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAF9B
Entrez Gene ID
102185255
Gene Name
TATA-box binding protein associated factor 9b
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 3439480 - 3448245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043790.1 XP_017899279.1 5 - 6 547 102
GenBank mRNA KA313326.1 - 5 - 6 474 102

View Full Product Details