Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYAA
Entrez Gene ID
102185956
Gene Name
crystallin alpha A
Gene Aliases
-
Chromosome Location
Chr.1: 142905036 - 142907851 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675651.3 XP_005675708.1 2 - 3 332 64

View Full Product Details