Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R3A
Entrez Gene ID
102183659
Gene Name
protein phosphatase 1 regulatory subunit 3A
Gene Aliases
-
Chromosome Location
Chr.4: 65654613 - 65694529 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679168.3 XP_005679225.2 2 - 3 862 105
XM_013963316.2 XP_013818770.2 3 - 4 551 105

View Full Product Details