Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYAR
Entrez Gene ID
102191413
Gene Name
Ly1 antibody reactive
Gene Aliases
-
Chromosome Location
Chr.6: 105125213 - 105143809 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681896.3 XP_005681953.2 4 - 5 548 88
GenBank mRNA JO576916.1 - 4 - 5 627 88
KA350157.1 - 5 - 6 916 88

View Full Product Details