Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHURC1
Entrez Gene ID
102188505
Gene Name
churchill domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 26059900 - 26077706 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685980.3 XP_005686037.1 2 - 3 278 80

View Full Product Details