Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAIP2B
Entrez Gene ID
102184251
Gene Name
poly(A) binding protein interacting protein 2B
Gene Aliases
-
Chromosome Location
Chr.11: 13218946 - 13262651 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686379.3 XP_005686436.2 2 - 3 329 84
GenBank mRNA JO450851.2 - 2 - 3 330 84
KA316957.1 - 2 - 3 326 84

View Full Product Details