Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL11B
Entrez Gene ID
102172563
Gene Name
methyltransferase like 11B
Gene Aliases
-
Chromosome Location
Chr.16: 36311290 - 36330052 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690631.3 XP_005690688.2 3 - 4 659 85

View Full Product Details