Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- THOC6
- Entrez Gene ID
-
102188727
- Gene Name
- THO complex 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.25: 2321861 - 2324788 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KA316702.1 |
- |
2 - 3 |
511 |
80 |
- Gene Symbol
- HCFC1R1
- Entrez Gene ID
-
100861053
- Gene Name
- host cell factor C1 regulator 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.25: 2320365 - 2322040 on Build ARS1
View Full Product Details