Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN16
Entrez Gene ID
102189431
Gene Name
claudin 16
Gene Aliases
-
Chromosome Location
Chr.1: 76623424 - 76647769 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963838.2 XP_013819292.2 2 - 3 552 72
GenBank mRNA JO437440.2 - 3 - 4 709 72

View Full Product Details