Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVOL3
Entrez Gene ID
102172926
Gene Name
ovo like zinc finger 3
Gene Aliases
-
Chromosome Location
Chr.18: 47612497 - 47615122 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971223.2 XP_013826677.1 2 - 3 852 98

View Full Product Details