Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102180889
Entrez Gene ID
102180889
Gene Name
transmembrane protein 45A
Gene Aliases
TMEM45A
Chromosome Location
Chr.1: 44694141 - 44805886 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO516519.2 - 1 - 2 157 98

View Full Product Details