Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF20
Entrez Gene ID
102168218
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
-
Chromosome Location
Chr.2: 20199137 - 20209976 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676635.2 XP_005676692.1 3 - 4 450 86

View Full Product Details