Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMF
Entrez Gene ID
100860972
Gene Name
Bcl2 modifying factor
Gene Aliases
-
Chromosome Location
Chr.10: 67305278 - 67326153 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013966966.2 XP_013822420.2 2 - 3 2561 66
XM_018054464.1 XP_017909953.1 4 - 5 1369 66
XM_018054465.1 XP_017909954.1 4 - 5 717 66
XM_018054466.1 XP_017909955.1 4 - 5 1070 66
XM_018054467.1 XP_017909956.1 4 - 5 1368 66
GenBank mRNA JI861610.1 - 2 - 3 446 66

View Full Product Details