The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005692127.3 | XP_005692184.2 | 3 - 4 | 543 | 82 |
XM_018062109.1 | XP_017917598.1 | 4 - 5 | 1543 | 82 | |
XM_018062110.1 | XP_017917599.1 | 3 - 4 | 996 | 82 | |
GenBank mRNA | JO576941.1 | - | 2 - 3 | 1086 | 82 |