Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C22H3orf14
Entrez Gene ID
102180896
Gene Name
chromosome 22 C3orf14 homolog
Gene Aliases
-
Chromosome Location
Chr.22: 38954086 - 38968985 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695765.3 XP_005695822.1 4 - 5 1108 97
XM_005695766.3 XP_005695823.1 4 - 5 1105 97

View Full Product Details