Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IQSEC3
Entrez Gene ID
102171957
Gene Name
IQ motif and Sec7 domain 3
Gene Aliases
-
Chromosome Location
Chr.5: 105971263 - 106060827 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048643.1 XP_017904132.1 4 - 5 2071 74
XM_018048644.1 XP_017904133.1 4 - 5 2071 74
XM_018048645.1 XP_017904134.1 4 - 5 2071 74
XM_018048646.1 XP_017904135.1 4 - 5 2071 74
XM_018048647.1 XP_017904136.1 2 - 3 1195 74
GenBank mRNA JO621633.2 - 3 - 4 1853 74
JO621634.2 - 3 - 4 1853 74

View Full Product Details