Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102178292
Entrez Gene ID
102178292
Gene Name
membrane-spanning 4-domains subfamily A member 5
Gene Aliases
MS4A5
Chromosome Location
Chr.15: 255590 - 281961 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058937.1 XP_017914426.1 1 - 2 248 71

View Full Product Details