Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOMER2
Entrez Gene ID
102178113
Gene Name
homer scaffold protein 2
Gene Aliases
-
Chromosome Location
Chr.21: 22406796 - 22532972 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972981.2 XP_013828435.1 3 - 4 617 90
XM_018066348.1 XP_017921837.1 3 - 4 491 90
GenBank mRNA KA336551.1 - 6 - 7 1758 90

View Full Product Details